6-Chloronicotinic acid
Glass bottle for solids, 1 g
MAYBSB01195.1
: 6-Chloropyridine-3-carboxylic acid, 6-Chloro-3-pyridinecarboxylic acid
- :5326-23-8
- :C₆H₄ClNO₂
- :MFCD00006241
- :157.56 g/mol
- :284 °C (1013 hPa)
- :188…189 °C
- :Ambient
- :226-201-5
- Pack type:Glass bottle for solids
- Pk:1 g