2-Methoxybenzylamine
Glass bottle for solids, 1 g
Catalog # MAYBSB01158.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: (2-Methoxyphenyl)methanamine, 2-(Aminomethyl)anisole, o-Anisylamine
- CAS Number:6850-57-3
- Formula:C₈H₁₁NO
- MDL Number:MFCD00008110
- Molecular Weight:137.18 g/mol
- Boiling Pt:227 °C (1013 hPa)
- Density:1.05 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:229-941-7
- UN:2735
- ADR:8,III
- :Density: 1,05 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



