2-Cyanopyrazine
Glass bottle for solids, 1 g
Catalog # MAYBSB01106.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile
- CAS Number:19847-12-2
- Formula:C₅H₃N₃
- MDL Number:MFCD00049361
- Molecular Weight:105.10 g/mol
- EINECS:243-369-5
- UN:3276
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



