1,3-Benzodioxole
Glass bottle for solids, 1 g
Catalog # MAYBSB00944.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: Benzo[d][1,3]dioxole, 1,2-Methylenedioxybenzene
- CAS Number:274-09-9
- Formula:C₇H₆O₂
- MDL Number:MFCD00005818
- Molecular Weight:122.12 g/mol
- Boiling Pt:172 °C (1013 hPa)
- Melting Pt:–18 °C
- Density:1,18 g/cm³ (20 °C)
- EINECS:205-992-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



