3,5-Dimethoxyaniline
Glass bottle for solids, 1 g
Catalog # MAYBSB00908.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:10272-07-8
- Formula:C₈H₁₁NO₂
- MDL Number:MFCD00008392
- Molecular Weight:153.18 g/mol
- Boiling Pt:303 °C (1013 hPa)
- Melting Pt:66…68 °C
- Storage Temperature:Ambient
- EINECS:233-616-5
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



