2,4-Difluorophenol
Glass bottle for solids, 10 g
Catalog # MAYBSB00632.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:367-27-1
- Formula:C₆H₄F₂O
- MDL Number:MFCD00009715
- Molecular Weight:130.09 g/mol
- Boiling Pt:146 °C (1013 hPa)
- Melting Pt:20 °C
- Density:1.36 g/cm³ (20 °C)
- EINECS:206-688-0
- UN:2921
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



