2,5-Dimethyl-2,5-hexanediol
Glass bottle for solids, 1 g
MAYBSB00569.1
: 1,1,4,4-Tetramethyl-1,4-butanediol
- :110-03-2
- :C₈H₁₈O₂
- :MFCD00004473
- :146.23 g/mol
- :214 °C (1013 hPa)
- :85…88 °C
- :0,898 g/cm³ (20 °C)
- :Ambient
- :203-731-5
- Pack type:Glass bottle for solids
- Pk:1 g