2,5-Dimethyl-2,5-hexanediol
Glass bottle for solids, 250 mg
Catalog # CHMPSB00569.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: 1,1,4,4-Tetramethyl-1,4-butanediol
- CAS Number:110-03-2
- Formula:C₈H₁₈O₂
- MDL Number:MFCD00004473
- Molecular Weight:146.23 g/mol
- Boiling Pt:214 °C (1013 hPa)
- Melting Pt:85…88 °C
- Density:0,898 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:203-731-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



