2,4-Difluoroaniline
Glass bottle for solids, 50 g
Catalog # MAYBSB00555.50
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:367-25-9
- Formula:C₆H₅F₂N
- MDL Number:MFCD00007648
- Molecular Weight:129.11 g/mol
- Merck Index:13,03171
- Boiling Pt:170 °C (1013 hPa)
- Melting Pt:–7 °C
- Density:1.268 g/cm³ (20 °C)
- Flash Pt:62 °C
- Storage Temperature:Ambient
- EINECS:206-687-5
- UN:2941
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



