4-Iodophenyl isothiocyanate
Glass bottle for solids, 50 g
MAYBS00350.50
: 1-Iodo-4-isothiocyanatobenzene, 4-Iodophenylisothiocyanate
- :2059-76-9
- :C₇H₄INS
- :MFCD00041091
- :261.09 g/mol
- :296 °C (1013 hPa)
- :75…78 °C
- :2923
- :8,III
- Pack type:Glass bottle for solids
- Pk:50 g