4-Iodophenyl isothiocyanate
Glass bottle for solids, 10 g
Catalog # MAYBS00350EA
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Iodo-4-isothiocyanatobenzene, 4-Iodophenylisothiocyanate
- CAS Number:2059-76-9
- Formula:C₇H₄INS
- MDL Number:MFCD00041091
- Molecular Weight:261.09 g/mol
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:75…78 °C
- UN:2923
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



