6-Fluoroindole
Glass bottle for solids, 250 mg
Catalog # CHMPRH00222.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: 6-Fluoro-1H-indole
- CAS Number:399-51-9
- Formula:C₈H₆FN
- MDL Number:MFCD00056933
- Molecular Weight:135.14 g/mol
- Melting Pt:72…77 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



