3-Ethoxybenzaldehyde
Glass bottle for solids, 10 g
Catalog # MAYBRH00790.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:22924-15-8
- Formula:C₉H₁₀O₂
- MDL Number:MFCD00016606
- Molecular Weight:150.18 g/mol
- Boiling Pt:243 °C (1013 hPa)
- Melting Pt:21 °C
- Density:1.06 g/cm³ (20 °C)
- EINECS:245-333-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



