4-Iodopyridine
Glass bottle for solids, 250 mg
Catalog # CHMPMO00014.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
- CAS Number:15854-87-2
- Formula:C₅H₄IN
- MDL Number:MFCD02093937
- Molecular Weight:205 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



