4-Nitroimidazole
Glass bottle for solids, 250 mg
Catalog # CHMPKM01643.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: 4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole
- CAS Number:3034-38-6
- Formula:C₃H₃N₃O₂
- MDL Number:MFCD00005196
- Molecular Weight:113.08 g/mol
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:221-224-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



