4-Chlorothioanisole
Glass bottle for solids, 10 g
Catalog # MAYBKM02761.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide
- CAS Number:123-09-1
- Formula:C₇H₇ClS
- MDL Number:MFCD00013643
- Molecular Weight:158.65 g/mol
- Boiling Pt:44 °C (0,08 mmHg)
- Boiling Pt:44 °C (0.08 mmHg)
- Melting Pt:27…28 °C
- Density:1,2151 g/cm³ (19 °C)
- EINECS:204-600-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



