4-Chlorothioanisole
Glass bottle for solids, 1 g
MAYBKM02761.1
: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide
- :123-09-1
- :C₇H₇ClS
- :MFCD00013643
- :158.65 g/mol
- :44 °C (0,08 mmHg)
- :44 °C (0.08 mmHg)
- :27…28 °C
- :1,2151 g/cm³ (19 °C)
- :204-600-5
- Pack type:Glass bottle for solids
- Pk:1 g