1-Phenylimidazole
Glass bottle for solids, 250 mg
Catalog # CHMPJFD01257.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
- CAS Number:7164-98-9
- Formula:C₉H₈N₂
- MDL Number:MFCD00041204
- Molecular Weight:144.18 g/mol
- Boiling Pt:276 °C (1013 hPa)
- Melting Pt:13 °C
- Density:1.1397 g/cm³ (20 °C)
- Storage Temperature:Ambient
- UN:2384
- ADR:3,II
- :Density: 1,1397 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



