2-Fluoro-m-xylene
Glass bottle for solids, 50 g
Catalog # MAYBCD05584.50
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Fluoro-1,3-dimethylbenzene, 1-Fluoro-2,6-dimethylbenzene
- CAS Number:443-88-9
- Formula:C₈H₉F
- MDL Number:MFCD00039215
- Molecular Weight:124.16 g/mol
- Boiling Pt:165…167 °C (1013 hPa)
- Density:0.988 g/cm³ (20 °C)
- EINECS:207-144-5
- UN:1993
- ADR:3,III
- :Density: 0.988 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



