5-Chloro-2-hydroxybenzonitrile
Glass bottle for solids, 1 g
Catalog # MAYBFM00044DA
Supplier: Thermo Scientific
5-Chloro-2-hydroxybenzonitrile
Catalog # MAYBFM00044DA
Supplier: Thermo Scientific
CAS Number: 13589-72-5
Jump to:
Product Details & Documents
- CAS Number:13589-72-5
- Formula:C₇H₄ClNO
- MDL Number:MFCD00219003
- Molecular Weight:153.5700 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



