2,6-Dimethoxybenzaldehyde
Glass bottle for solids, 10 g
Catalog # MAYBCD11827.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:3392-97-0
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00010862
- Molecular Weight:166.18 g/mol
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:95…96 °C
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



