2-Bromopyridine
Glass bottle for solids, 10 g
Catalog # MAYBCC04010.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:109-04-6
- Formula:C₅H₄BrN
- MDL Number:MFCD00006219
- Molecular Weight:158 g/mol
- Boiling Pt:192 °C (1013 hPa)
- Melting Pt:–40.1 °C
- Density:1.62 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:203-641-6
- UN:2810
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



