4-Bromo-3,5-dimethyl-1H-pyrazole
Glass bottle for solids, 10 g
Catalog # MAYBCC01210.10
Supplier: Thermo Scientific
4-Bromo-3,5-dimethyl-1H-pyrazole
Catalog # MAYBCC01210.10
Supplier: Thermo Scientific
CAS Number: 3398-16-1
Jump to:
Product Details & Documents
Synonyms: 4-Bromo-3,5-dimethylpyrazole
- CAS Number:3398-16-1
- Formula:C₅H₇BrN₂
- MDL Number:MFCD00005242
- Molecular Weight:175.03 g/mol
- EINECS:222-263-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



