1,2,3,4,6,7-Hexabromonaphthalene
Glass bottle for solids, 250 mg
Catalog # MAYBBTB15044.250
Supplier: Thermo Scientific
1,2,3,4,6,7-Hexabromonaphthalene
Catalog # MAYBBTB15044.250
Supplier: Thermo Scientific
CAS Number: 75625-24-0
Jump to:
Product Details & Documents
- CAS Number:75625-24-0
- Formula:C₁₀H₂Br₆
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



