Order Entry
Export
Orders LinkContactUsLinkComponent
4-Methoxybenzonitrile

4-Methoxybenzonitrile

Glass bottle for solids, 1 g
Catalog # MAYBCD00358.1
Irritant
Supplier:   Thermo Scientific
4-Methoxybenzonitrile
Catalog # MAYBCD00358.1
Supplier:  Thermo Scientific
CAS Number:  874-90-8
4-Methoxybenzonitrile
4-Methoxybenzonitrile

Glass bottle for solids, 1 g

CAS Number:  874-90-8
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
  • CAS Number:874-90-8
  • Formula:C₈H₇NO
  • MDL Number:MFCD00001818
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:256 °C (1013 hPa)
  • Melting Pt:57…60 °C
  • Storage Temperature:Ambient
  • EINECS:212-871-6
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications