2-Thiophenemethanol
Glass bottle for solids, 50 g
Catalog # MAYBCC13009.50
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: Thiophen-2-methanol, Thiophen-2-ylmethanol, 2-Thienylmethanol
- CAS Number:636-72-6
- Formula:C₅H₆OS
- MDL Number:MFCD00005454
- Molecular Weight:114.17 g/mol
- Boiling Pt:207 °C (1013 hPa)
- Melting Pt:0 °C
- Density:1.206 g/cm³ (20 °C)
- EINECS:211-264-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



