3-(Hydroxymethyl)indole
Glass bottle for solids, 1 g
Catalog # MAYBCC12709.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol
- CAS Number:700-06-1
- Formula:C₉H₉NO
- MDL Number:MFCD00005632
- Molecular Weight:147.18 g/mol
- EINECS:211-836-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



