2'-Hydroxy-4'-methoxyacetophenone
Glass bottle for solids, 10 g
Catalog # MAYBBTB09987.10
Supplier: Thermo Scientific
2'-Hydroxy-4'-methoxyacetophenone
Catalog # MAYBBTB09987.10
Supplier: Thermo Scientific
CAS Number: 552-41-0
Jump to:
Product Details & Documents
Synonyms: 1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
- CAS Number:552-41-0
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00008730
- Molecular Weight:166.18 g/mol
- Boiling Pt:154 °C (20 mmHg)
- Melting Pt:51 °C
- Density:1,1604 g/cm³ (40 °C)
- EINECS:209-012-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



