Diphenylcyclopropenone
Glass bottle for solids, 1 g
Catalog # MAYBBTB10303.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one
- CAS Number:886-38-4
- Formula:C₁₅H₁₀O
- MDL Number:MFCD00001311
- Molecular Weight:206.24 g/mol
- Merck Index:12,03366
- Boiling Pt:407 °C (1013 hPa)
- Melting Pt:112…113 °C
- Density:1.1 g/cm³ (20 °C)
- EINECS:212-948-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



