6-Bromochromone
Glass bottle for solids, 1 g
Catalog # MAYBBTB14078.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 6-Bromo-4H-chromen-4-one
- CAS Number:51483-92-2
- Formula:C₉H₅BrO₂
- MDL Number:MFCD00239369
- Molecular Weight:225.0388 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



