6-Bromochromone
Glass bottle for solids, 250 mg
Catalog # CHMPBTB14078.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: 6-Bromo-4H-chromen-4-one
- CAS Number:51483-92-2
- Formula:C₉H₅BrO₂
- MDL Number:MFCD00239369
- Molecular Weight:225.0388 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



