4'-Hexylacetophenone
10 g
Catalog # MAYBBTB09786.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: p-Hexylacetophenone, 1-(4-Hexylphenyl)ethanone
- CAS Number:37592-72-6
- Formula:C₁₄H₂₀O
- MDL Number:MFCD00043677
- Molecular Weight:204.31
Specifications
Specifications
- Pk:10 g



