Order Entry
Export
Orders LinkContactUsLinkComponent
ω-Bromo-4-fluoroacetophenone

ω-Bromo-4-fluoroacetophenone

Glass bottle for solids, 50 g
Catalog # MAYBBTB05530.50
Supplier:   Thermo Scientific
ω-Bromo-4-fluoroacetophenone
Catalog # MAYBBTB05530.50
Supplier:  Thermo Scientific
CAS Number:  403-29-2
ω-Bromo-4-fluoroacetophenone

Glass bottle for solids, 50 g

CAS Number:  403-29-2
Price
Inquire For Price
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone
  • CAS Number:403-29-2
  • Formula:C₈H₆BrFO
  • MDL Number:MFCD00040830
  • Molecular Weight:217.04 g/mol
  • Boiling Pt:82 °C (0,4 mmHg)
  • Boiling Pt:82 °C (0.4 mmHg)
  • Melting Pt:49 °C
  • EINECS:206-955-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Cat. No.:
    MAYBBTB05530.50

Specifications