2-(Ethylthio)nicotinic acid
Glass bottle for solids, 10 g
MAYBBTB06379EA
Jump to:
- CAS Number:27868-76-4
- Formula:C₈H₉NO₂S
- MDL Number:MFCD00051680
- Molecular Weight:183.23 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



