2-(Ethylthio)nicotinic acid
Glass bottle for solids, 1 g
Catalog # MAYBBTB06379DA
Supplier: Thermo Scientific
2-(Ethylthio)nicotinic acid
Catalog # MAYBBTB06379DA
Supplier: Thermo Scientific
CAS Number: 27868-76-4
Jump to:
Product Details & Documents
- CAS Number:27868-76-4
- Formula:C₈H₉NO₂S
- MDL Number:MFCD00051680
- Molecular Weight:183.23 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



