4-Bromo-2,6-xylidine
Glass bottle for solids, 10 g
Catalog # MAYBBTB04886.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 4-Bromo-2,6-dimethylaniline
- CAS Number:24596-19-8
- Formula:C₈H₁₀BrN
- MDL Number:MFCD00007826
- Molecular Weight:200.08 g/mol
- Boiling Pt:120 °C (0.2 mmHg)
- Melting Pt:50 °C
- EINECS:246-337-9
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



