2-(4-Chlorophenoxy)nicotinic acid
Glass bottle for solids, 50 g
Catalog # MAYBBTB08733.50
Supplier: Thermo Scientific
2-(4-Chlorophenoxy)nicotinic acid
Catalog # MAYBBTB08733.50
Supplier: Thermo Scientific
CAS Number: 51362-37-9
Jump to:
Product Details & Documents
- CAS Number:51362-37-9
- Formula:C₁₂H₈ClNO₃
- MDL Number:MFCD00052116
- Molecular Weight:249.64 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



