2,6-Dichlorophenylhydrazinium chloride
Glass bottle for solids, 1 g
Catalog # MAYBBTB08663DA
Supplier: Thermo Scientific
2,6-Dichlorophenylhydrazinium chloride
Catalog # MAYBBTB08663DA
Supplier: Thermo Scientific
CAS Number: 50709-36-9
Jump to:
Product Details & Documents
Synonyms: 2,6-Dichlorophenylhydrazine hydrochloride, 1-(2,6-Dichlorophenyl)hydrazine hydrochloride
- CAS Number:50709-36-9
- Formula:C₆H₇Cl₃N₂
- MDL Number:MFCD00012930
- Molecular Weight:213.49 g/mol
- EINECS:256-730-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



