5-Hydroxy-DL-tryptophan
Glass bottle for solids, 10 g
Catalog # MAYBAC12199.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: DL-5-Hydroxytryptophan, 2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
- CAS Number:56-69-9
- Formula:C₁₁H₁₂N₂O₃
- MDL Number:MFCD00005651
- Molecular Weight:220.23 g/mol
- Merck Index:13,04873
- Melting Pt:301 °C
- Density:1.405 g/cm³ (20 °C)
- EINECS:200-284-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



