2,6-Dichloropyrazine 99+%
Glass bottle for solids, 10 g
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:4774-14-5
- Formula:C₄H₂Cl₂N₂
- MDL Number:MFCD00006125
- Molecular Weight:148.98 g/mol
- Boiling Pt:120 - 122 °C (40 -rr)
- Melting Pt:53…56 °C
- Density:1,713 g/cm³ (20 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:225-316-8
Documents
ACRO23254
SDS
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:53 to 56 °C
- GC:≥99.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



