7-Methylindole
Glass bottle for solids, 10 g
MAYBAC12704.10
- :933-67-5
- :C₉H₉N
- :MFCD00005684
- :131.18 g/mol
- :266 °C (1013 hPa)
- :83 °C
- :1.0202 g/cm³ (99.6 °C)
- :Density: 1.0202 g/cm³ (99.6 °C)
- :213-270-1
- Pack type:Glass bottle for solids
- Pk:10 g
- Cat. No.:MAYBAC12704.10