0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
7-Methylindole
7-Methylindole
7-Methylindole
Glass bottle for solids, 1 g
MAYBAC12704.1
:
Thermo Scientific
7-Methylindole
MAYBAC12704.1
:
Thermo Scientific
:
933-67-5
7-Methylindole
Glass bottle for solids, 1 g
:
933-67-5
1 * 1 g
-
+
:
933-67-5
:
C₉H₉N
:
MFCD00005684
:
131.18 g/mol
:
266 °C (1013 hPa)
:
83 °C
:
1.0202 g/cm³ (99.6 °C)
:
Density: 1.0202 g/cm³ (99.6 °C)
:
213-270-1
Pack type:
Glass bottle for solids
Pk:
1 g