2,4-Dichlorophenyl acetate
Glass bottle for solids, 250 mg
Catalog # CHMPBLT00260.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
- CAS Number:6341-97-5
- Formula:C₈H₆Cl₂O₂
- Molecular Weight:205.0380 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



