Proteinase K
Glass bottle, 6,25 mL
Catalog # AMBI2548
Supplier: Ambion
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:6,25 mL
- Cat. No.:AMBI2548
- Enzyme activity:600 mAnsonU/ml
- Form:Aqueous solution
- Storage conditions:Refrigerator
- Concentration:20 mg/ml
- Enzyme name:Proteinase K
- CAS No.:39450-01-6



