Proteinase K
Glass bottle for solids, 500 mg
Catalog # AMBI2544
Supplier: Ambion
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Cat. No.:AMBI2544
- Storage conditions:Freezer
- Enzyme name:Proteinase K
- CAS No.:39450-01-6



