Order Entry
Export
Orders LinkContactUsLinkComponent
Proteinase K

Proteinase K

Glass bottle for solids, 100 mg
Catalog # AMBI2542
Supplier:   Ambion
Proteinase K
Catalog # AMBI2542
Supplier:  Ambion
CAS Number:  
Proteinase K

Glass bottle for solids, 100 mg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Cat. No.:
    AMBI2542
  • Storage conditions:
    Freezer
  • Enzyme name:
    Proteinase K
  • CAS No.:
    39450-01-6

Specifications