Buffer, SSC solution 20X concentrate (saline-sodium citrate)
SSC buffer solution 20X concentrate (saline-sodium citrate), Glass bottle, 500 mL
Catalog # AMBI9770
Supplier: Ambion
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # AMBI9770
Supplier: Ambion
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:SSC buffer solution 20X concentrate (saline-sodium citrate)



