0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Buffers
Electrophoresis Buffers
Hybridisation Buffers
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
SSC buffer solution 20X concentrate (saline-sodium citrate), 4 L
AMBI9765
:
Ambion
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
AMBI9765
:
Ambion
:
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
SSC buffer solution 20X concentrate (saline-sodium citrate), 4 L
:
1 * 4 L
-
+
Pk:
4 L
Description:
SSC buffer solution 20X concentrate (saline-sodium citrate)