Buffer, SSC solution 20X concentrate (saline-sodium citrate)
SSC buffer solution 20X concentrate (saline-sodium citrate), 1 KIT
Catalog # AMBI9764
Supplier: Ambion
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # AMBI9764
Supplier: Ambion
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 KIT
- Description:SSC buffer solution 20X concentrate (saline-sodium citrate)



