Ammonium acetate in aqueous solution
Glass bottle, 1 mL
Catalog # AMBI9077G
Supplier: Ambion
Jump to:
Product Details & Documents
Synonyms: Acetic acid ammonium salt
- CAS Number:631-61-8
- Formula:H₃CCOONH₄
- MDL Number:MFCD00013066
- Molecular Weight:77.08 g/mol
- Storage Temperature:Ambient
- EINECS:211-162-9
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 mL



